Assay Details
Gene Symbol: Pitpnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024927.2 | NP_001020098.1 | 14 - 15 | 2100 | 72 | |
NM_001081641.1 | NP_001075110.1 | 13 - 14 | 2052 | 72 | ||
GenBank mRNA | AK147444.1 | - | 14 - 15 | 2057 | 72 | |
AK147630.1 | - | 13 - 14 | 1885 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
