Assay Details
Gene Symbol: Cxcl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023158.6 | NP_075647.3 | 1 - 2 | 604 | 69 | |
GenBank mRNA | AF277001.1 | - | 1 - 2 | 67 | 69 | |
AF301017.1 | - | 1 - 2 | 133 | 69 | ||
AK003085.1 | - | 1 - 2 | 264 | 69 | ||
AK009599.1 | - | 1 - 2 | 153 | 69 | ||
AK028875.1 | - | 1 - 2 | 606 | 69 | ||
AK150224.1 | - | 1 - 2 | 292 | 69 | ||
AK151330.1 | - | 1 - 2 | 378 | 69 | ||
AK152102.1 | - | 1 - 2 | 378 | 69 | ||
AK152547.1 | - | 1 - 2 | 556 | 69 | ||
AK153192.1 | - | 1 - 2 | 320 | 69 | ||
AK170428.1 | - | 1 - 2 | 453 | 69 | ||
BC019961.1 | - | 1 - 2 | 485 | 3988542 | 69 | |
GQ380016.1 | - | 1 - 2 | 100 | 69 | ||
GQ380017.1 | - | 1 - 2 | 100 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
