Assay Details
Gene Symbol: BC100451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021440.2 | NP_067415.2 | 2 - 3 | 320 | 56 | |
GenBank mRNA | AK132657.1 | - | 2 - 3 | 128 | 56 | |
BC100451.1 | - | 2 - 3 | 320 | 30918483 | 56 | |
Y09878.1 | - | 2 - 3 | 286 | 56 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK132657.1, BC100451.1, NM_021440.2, Y09878.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | BC100451.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | BX520604.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | Y09878.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | BC100451.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | BX520604.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | Y09878.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151732 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140072 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137327 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | BC100451.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | BX520604.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | Y09878.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | AK132657.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | BC100451.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | BX520604.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | Y09878.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | BC100451.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | BX520604.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | Y09878.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | BC100451.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | BX520604.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | Y09878.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170737 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170726 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28170769 | - | - |
Whole Mouse Genome G4122A | A_51_P441494 | BC100451.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | BX520604.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | Y09878.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | BC100451.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | BX520604.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | Y09878.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | NM_021440.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P441494 | AK132657.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111934 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09457630 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09457624 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09457623 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09457637 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09457635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28170749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08980111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09398814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09149419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09398823 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09503784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09687733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09149429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09431996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09398802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09398804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09355089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09149417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08993208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08980115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09503785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09355095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28170720 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10393571 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393570 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393572 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450469_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10393569 | - | - |
Mouse Genome 430 2.0 Array | 1450469_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450469_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58251_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58251_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100039529.mm.1 | TC1100004158.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100039529.mm.1 | NM_021440.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100039529.mm.1 | NM_021440.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100039528.mm.1 | TC1100004158.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100039528.mm.1 | NM_021440.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100039528.mm.1 | NM_021440.2 | Refseq |
Mouse Expression Set 430 | 1450469_PM_at | - | - |
Mouse Expression Set 430 | 1450469_at | - | - |
Murine Genome U74v2 Set | 92455_at | - | - |
Murine Genome U74v2 Set | 92455_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450469_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450469_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450469_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450469_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 353977 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6650019 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6650019 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006650019 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248326 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248326 | - | - |
mouse-6 v1.1 (Illumina) | scl39276.3_603-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6650019 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39276.3_603-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650019 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1248326 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248326 | - | - |
