Assay Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU887626.1 | - | 1 - 2 | 72 | 131 | |
EU887627.1 | - | 1 - 2 | 72 | 131 | ||
EU887628.1 | - | 1 - 2 | 72 | 131 | ||
U28807.1 | - | 1 - 2 | 63 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
