Assay Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC021835.1 | - | 6 - 7 | 1597 | 68 | |
EU887626.1 | - | 9 - 10 | 3081 | 68 | ||
EU887629.1 | - | 9 - 10 | 3105 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
