Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019825.3 | NP_062799.2 | 9 - 10 | 3071 | 64 | |
GenBank mRNA | AF135169.1 | - | 2 - 3 | 569 | 64 | |
AF216186.1 | - | 8 - 9 | 2956 | 64 | ||
AK129079.1 | - | 2 - 3 | 230 | 64 | ||
AK142566.1 | - | 3 - 4 | 1256 | 64 | ||
AK171864.1 | - | 9 - 10 | 3229 | 64 | ||
BC083187.1 | - | 9 - 10 | 3071 | 30544739 | 64 | |
JF707635.1 | - | 8 - 9 | 2930 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
