Assay Details
Gene Symbol: Pdcd6ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164677.1 | NP_001158149.1 | 10 - 11 | 1532 | 75 | |
NM_001164678.1 | NP_001158150.1 | 10 - 11 | 1517 | 75 | ||
NM_011052.2 | NP_035182.2 | 10 - 11 | 1517 | 75 | ||
GenBank mRNA | AF119955.1 | - | 10 - 11 | 1478 | 75 | |
AJ005073.1 | - | 10 - 11 | 1365 | 75 | ||
AK029827.1 | - | 10 - 11 | 1484 | 75 | ||
AK031256.1 | - | 10 - 11 | 1518 | 75 | ||
AK033943.1 | - | 10 - 11 | 1506 | 75 | ||
AK145893.1 | - | 10 - 11 | 1505 | 75 | ||
AK147736.1 | - | 10 - 11 | 1490 | 75 | ||
AK167574.1 | - | 10 - 11 | 1475 | 75 | ||
AK169704.1 | - | 10 - 11 | 1518 | 75 | ||
BC026823.1 | - | 10 - 11 | 1459 | 4038684 | 75 | |
BC051123.1 | - | 10 - 11 | 1368 | 6529850 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
