Assay Details
Gene Symbol: Tcf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286362.1 | NP_001273291.1 | 16 - 17 | 1963 | 75 | |
NM_025804.3 | NP_080080.2 | 17 - 18 | 2038 | 75 | ||
GenBank mRNA | AK013754.1 | - | 17 - 18 | 1727 | 75 | |
AK045397.1 | - | 16 - 17 | 1892 | 75 | ||
AK050274.1 | - | 17 - 18 | 1964 | 75 | ||
AK075728.1 | - | 17 - 18 | 1888 | 75 | ||
AK088602.1 | - | 4 - 5 | 577 | 75 | ||
BC046768.1 | - | 17 - 18 | 1996 | 5707312 | 75 | |
U94988.1 | - | 17 - 18 | 1899 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
