Assay Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009603.1 | NP_033733.1 | 4 - 5 | 367 | 61 | |
GenBank mRNA | X55718.1 | - | 4 - 5 | 367 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
