Assay Details
Gene Symbol: Prmt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145404.1 | NP_663379.1 | 7 - 8 | 872 | 73 | |
GenBank mRNA | AK154255.1 | - | 7 - 8 | 899 | 73 | |
AK173304.1 | - | 7 - 8 | 892 | 73 | ||
AY673972.1 | - | 6 - 7 | 742 | 73 | ||
BC006705.1 | - | 7 - 8 | 872 | 3583763 | 73 | |
BC012845.1 | - | 6 - 7 | 693 | 73 | ||
BC057177.1 | - | 7 - 8 | 896 | 5010547 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
