Assay Details
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010575.2 | NP_034705.2 | 11 - 12 | 1166 | 77 | |
GenBank mRNA | AF166384.1 | - | 11 - 12 | 1044 | 77 | |
AF170316.1 | - | 11 - 12 | 1027 | 77 | ||
AK140749.1 | - | 6 - 7 | 818 | 77 | ||
AK142289.1 | - | 11 - 12 | 1028 | 77 | ||
AK154619.1 | - | 11 - 12 | 1167 | 77 | ||
AK156872.1 | - | 10 - 11 | 1348 | 77 | ||
AK157012.1 | - | 10 - 11 | 1349 | 77 | ||
AK158443.1 | - | 11 - 12 | 1013 | 77 | ||
BC120493.1 | - | 11 - 12 | 1016 | 40129416 | 77 | |
BC137570.1 | - | 11 - 12 | 1016 | 8860589 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
