Assay Details
Gene Symbol: Mpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007863.2 | NP_031889.2 | 5 - 6 | 487 | 85 | |
GenBank mRNA | AF079366.1 | - | 5 - 6 | 438 | 85 | |
AK032270.1 | - | 4 - 5 | 1761 | 85 | ||
AK047456.1 | - | 5 - 6 | 467 | 85 | ||
AK163205.1 | - | 5 - 6 | 470 | 85 | ||
AY236514.1 | - | 4 - 5 | 321 | 85 | ||
BC052003.1 | - | 734 | 85 | |||
BC062642.1 | - | 5 - 6 | 455 | 5718318 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
