Assay Details
Gene Symbol: Rbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139516.1 | NP_001132988.1 | 2 - 3 | 368 | 96 | |
NM_011249.2 | NP_035379.2 | 2 - 3 | 368 | 96 | ||
GenBank mRNA | AK033402.1 | - | 2 - 3 | 362 | 96 | |
AK147064.1 | - | 2 - 3 | 369 | 96 | ||
AK156055.1 | - | 2 - 3 | 347 | 96 | ||
BC060124.1 | - | 2 - 3 | 335 | 6408844 | 96 | |
U27177.1 | - | 2 - 3 | 352 | 96 | ||
U27178.1 | - | 2 - 3 | 352 | 96 | ||
U33320.1 | - | 2 - 3 | 335 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
