Assay Details
Gene Symbol: Aldh1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153543.2 | NP_705771.2 | 19 - 20 | 2368 | 93 | |
GenBank mRNA | AK052361.1 | - | 19 - 20 | 2259 | 93 | |
AK141954.1 | - | 19 - 20 | 2329 | 93 | ||
BC034531.1 | - | 19 - 20 | 2290 | 4188055 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
