Assay Details
Gene Symbol: Slc9a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081332.1 | NP_001074801.1 | 13 - 14 | 3146 | 60 | |
GenBank mRNA | AK134092.1 | - | 8 - 9 | 1467 | 60 | |
BC057616.1 | - | 9 - 10 | 1908 | 60 | ||
BC151197.1 | - | 13 - 14 | 2587 | 9088085 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
