Assay Details
Gene Symbol: Agrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271806.1 | NP_001258735.1 | 4 - 5 | 533 | 77 | |
NM_007427.3 | NP_031453.1 | 4 - 5 | 423 | 77 | ||
GenBank mRNA | BC079902.1 | - | 4 - 5 | 420 | 30611244 | 77 |
CO044337.1 | - | 1 - 2 | 77 | |||
U89484.1 | - | 2 - 3 | 213 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26134617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26134616 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049717 | BC079902.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049717 | AK138796.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049717 | NM_007427.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589386 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604056 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151352 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799310 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593662 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32072 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643297 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304795 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141102 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578133 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89572 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917749 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418454 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645138 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714521 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262494 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309473 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845838 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511219 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681819 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150713 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122102 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31339 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157442 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136873 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165993 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170747 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141623 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049717 | BC079902.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049717 | AK138796.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049717 | NM_007427.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P347965 | BC079902.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P347965 | U89484.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P347965 | AK138796.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P347965 | NM_007427.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134641 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134639 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134626 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141623 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06627031 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01818076 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170747 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06627016 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01818117 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136873 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06627003 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165993 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01818058 | - | - |
Whole Mouse Genome G4122A | A_51_P347965 | BC079902.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P347965 | U89484.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P347965 | AK138796.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P347965 | NM_007427.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125036 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112605 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01818058 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136873 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06627016 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170747 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165993 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141623 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07146712 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928381 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928375 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07146714 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928404 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928387 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928393 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07146713 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07146715 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26134617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26134616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06805666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07288710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06928392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06939295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07146712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07154450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07154451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06933374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06861690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06928398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07348798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07348796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26134672 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581302 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581303 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581304 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581301 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581300 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581305 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421690_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581299 | - | - |
Mouse Genome 430 2.0 Array | 1421690_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1421690_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11604_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11604_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800021758.mm.1 | TC0800003246.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800021758.mm.1 | NM_001271806.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021758.mm.1 | NM_007427.3 | Refseq |
Mouse Expression Set 430 | 1421690_s_at | - | - |
Mouse Expression Set 430 | 1421690_PM_s_at | - | - |
Murine Genome U74v2 Set | 99399_at | - | - |
Murine Genome U74v2 Set | 99399_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421690_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1421690_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421690_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421690_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 796850 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1990082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001990082 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2703427 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2703427 | - | - |
mouse-6 v1.1 (Illumina) | scl34398.4.1_108-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1990082 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl34398.4.1_108-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990082 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2703427 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2703427 | - | - |
