Assay Details
Gene Symbol: Lgr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010195.2 | NP_034325.2 | 14 - 15 | 1567 | 75 | |
GenBank mRNA | AF110818.1 | - | 14 - 15 | 1280 | 75 | |
AK132387.1 | - | 14 - 15 | 1567 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
