Assay Details
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016776.2 | NP_058056.2 | 1 - 2 | 196 | 69 | |
GenBank mRNA | AK083908.1 | - | 1 - 2 | 220 | 69 | |
AK143487.1 | - | 1 - 2 | 229 | 69 | ||
AK154016.1 | - | 1 - 2 | 218 | 69 | ||
AK155074.1 | - | 1 - 2 | 207 | 69 | ||
AK166533.1 | - | 1 - 2 | 220 | 69 | ||
AK208884.1 | - | 1 - 2 | 100 | 69 | ||
BC015070.1 | - | 1 - 2 | 197 | 69 | ||
BC048858.1 | - | 1 - 2 | 196 | 5363740 | 69 | |
BC052889.1 | - | 1 - 2 | 217 | 30057169 | 69 | |
BC060167.1 | - | 1 - 2 | 202 | 69 | ||
U63648.1 | - | 1 - 2 | 205 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
