Assay Details
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010578.2 | NP_034708.1 | 11 - 12 | 1773 | 79 | |
GenBank mRNA | AK076526.1 | - | 7 - 8 | 1070 | 79 | |
AK088016.1 | - | 11 - 12 | 1674 | 79 | ||
AK088729.1 | - | 11 - 12 | 1520 | 79 | ||
AK135521.1 | - | 10 - 11 | 1449 | 79 | ||
AK141253.1 | - | 6 - 7 | 700 | 79 | ||
AK159689.1 | - | 11 - 12 | 1615 | 79 | ||
AK167682.1 | - | 11 - 12 | 1671 | 79 | ||
BC050906.1 | - | 11 - 12 | 1646 | 5721348 | 79 | |
X15202.1 | - | 10 - 11 | 1469 | 79 | ||
Y00769.1 | - | 11 - 12 | 1561 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
