Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 12 - 13 | 1306 | 76 | |
NM_026653.2 | NP_080929.1 | 11 - 12 | 1243 | 76 | ||
GenBank mRNA | AK019864.1 | - | 5 - 6 | 644 | 76 | |
AK035084.1 | - | 11 - 12 | 1171 | 76 | ||
AK148035.1 | - | 11 - 12 | 1184 | 76 | ||
AK150785.1 | - | 11 - 12 | 1140 | 76 | ||
AK151590.1 | - | 11 - 12 | 1196 | 76 | ||
AK152296.1 | - | 12 - 13 | 1236 | 76 | ||
AK153107.1 | - | 12 - 13 | 1236 | 76 | ||
AK153144.1 | - | 11 - 12 | 1196 | 76 | ||
AK165316.1 | - | 11 - 12 | 1156 | 76 | ||
AK165944.1 | - | 11 - 12 | 1174 | 76 | ||
AK167598.1 | - | 11 - 12 | 1129 | 76 | ||
AK168400.1 | - | 11 - 12 | 1196 | 76 | ||
AK169599.1 | - | 11 - 12 | 1167 | 76 | ||
AK194307.1 | - | 2 - 3 | 172 | 76 | ||
AK194339.1 | - | 2 - 3 | 283 | 76 | ||
AK196134.1 | - | 2 - 3 | 170 | 76 | ||
AK201262.1 | - | 2 - 3 | 282 | 76 | ||
AK203011.1 | - | 2 - 3 | 282 | 76 | ||
AK204019.1 | - | 2 - 3 | 282 | 76 | ||
AK204900.1 | - | 2 - 3 | 76 | |||
AK205283.1 | - | 1 - 2 | 138 | 76 | ||
AK210768.1 | - | 2 - 3 | 178 | 76 | ||
AK217077.1 | - | 2 - 3 | 282 | 76 | ||
AK219356.1 | - | 2 - 3 | 75 | |||
BC019119.1 | - | 11 - 12 | 1174 | 4487586 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
