Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 1 - 2 | 158 | 103 | |
NM_026653.2 | NP_080929.1 | 1 - 2 | 158 | 103 | ||
GenBank mRNA | AK035084.1 | - | 1 - 2 | 86 | 103 | |
AK148035.1 | - | 1 - 2 | 99 | 103 | ||
AK151590.1 | - | 1 - 2 | 111 | 103 | ||
AK152296.1 | - | 1 - 2 | 88 | 103 | ||
AK153107.1 | - | 1 - 2 | 88 | 103 | ||
AK153144.1 | - | 1 - 2 | 111 | 103 | ||
AK165316.1 | - | 1 - 2 | 71 | 103 | ||
AK165944.1 | - | 1 - 2 | 89 | 103 | ||
AK168400.1 | - | 1 - 2 | 107 | 103 | ||
AK169599.1 | - | 1 - 2 | 88 | 103 | ||
BC019119.1 | - | 1 - 2 | 89 | 4487586 | 103 | |
CJ078143.1 | - | 1 - 2 | 158 | 103 | ||
CJ086677.1 | - | 1 - 2 | 158 | 103 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK148035.1, AK151590.1, AK152296.1, AK153144.1, AK165316.1, AK165944.1, AK169599.1, BC019119.1, NM_001164223.1, NM_026653.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
