Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 3 - 4 | 290 | 72 | |
NM_026653.2 | NP_080929.1 | 3 - 4 | 290 | 72 | ||
GenBank mRNA | AK035084.1 | - | 3 - 4 | 218 | 72 | |
AK148035.1 | - | 3 - 4 | 231 | 72 | ||
AK150785.1 | - | 3 - 4 | 187 | 72 | ||
AK151590.1 | - | 3 - 4 | 243 | 72 | ||
AK152296.1 | - | 3 - 4 | 220 | 72 | ||
AK153107.1 | - | 3 - 4 | 220 | 72 | ||
AK153144.1 | - | 3 - 4 | 243 | 72 | ||
AK165316.1 | - | 3 - 4 | 203 | 72 | ||
AK165944.1 | - | 3 - 4 | 221 | 72 | ||
AK167598.1 | - | 3 - 4 | 182 | 72 | ||
AK168400.1 | - | 3 - 4 | 239 | 72 | ||
AK169599.1 | - | 3 - 4 | 220 | 72 | ||
AK180105.1 | - | 1 - 2 | 45 | 72 | ||
AK197958.1 | - | 3 - 4 | 175 | 72 | ||
AK199793.1 | - | 1 - 2 | 45 | 72 | ||
BC019119.1 | - | 3 - 4 | 221 | 4487586 | 72 | |
CJ078143.1 | - | 3 - 4 | 290 | 72 | ||
CJ086677.1 | - | 3 - 4 | 290 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
