Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 6 - 7 | 541 | 132 | |
NM_026653.2 | NP_080929.1 | 5 - 6 | 478 | 132 | ||
GenBank mRNA | AK035084.1 | - | 5 - 6 | 406 | 132 | |
AK148035.1 | - | 5 - 6 | 419 | 132 | ||
AK150785.1 | - | 5 - 6 | 375 | 132 | ||
AK151590.1 | - | 5 - 6 | 431 | 132 | ||
AK153144.1 | - | 5 - 6 | 431 | 132 | ||
AK165316.1 | - | 5 - 6 | 391 | 132 | ||
AK165944.1 | - | 5 - 6 | 409 | 132 | ||
AK167598.1 | - | 5 - 6 | 370 | 126 | ||
AK168400.1 | - | 5 - 6 | 427 | 135 | ||
AK169599.1 | - | 5 - 6 | 408 | 126 | ||
BC019119.1 | - | 5 - 6 | 409 | 4487586 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
