Assay Details
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177980.1 | NP_001171451.1 | 1 - 2 | 826 | 84 | |
NM_019840.2 | NP_062814.2 | 6 - 7 | 991 | 84 | ||
GenBank mRNA | AF208023.2 | - | 6 - 7 | 926 | 84 | |
AF326555.1 | - | 6 - 7 | 731 | 84 | ||
AF326556.1 | - | 6 - 7 | 731 | 84 | ||
AJ297397.1 | - | 6 - 7 | 926 | 84 | ||
AK150868.1 | - | 1 - 2 | 332 | 84 | ||
BC007155.1 | - | 1 - 2 | 173 | 84 | ||
BC023751.1 | - | 1 - 2 | 802 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
