Assay Details
Gene Symbol: Pgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163314.2 | NP_001156786.1 | 10 - 11 | 1275 | 158 | |
GenBank mRNA | AK041133.1 | - | 10 - 11 | 1276 | 158 | |
AK138171.1 | - | 10 - 11 | 1215 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
