Assay Details
Gene Symbol: Pgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163314.2 | NP_001156786.1 | 14 - 15 | 1534 | 100 | |
GenBank mRNA | AK138171.1 | - | 14 - 15 | 1474 | 100 | |
CK392830.1 | - | 1 - 2 | 68 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
