Assay Details
Gene Symbol: Nck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010879.3 | NP_035009.3 | 1 - 2 | 279 | 59 | |
GenBank mRNA | AK014949.1 | - | 1 - 2 | 272 | 59 | |
AK051665.1 | - | 1 - 2 | 279 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK051665.1, NM_010879.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
