Assay Details
Gene Symbol: Taf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100449.1 | NP_001093919.1 | 14 - 15 | 2756 | 73 | |
GenBank mRNA | AK029943.1 | - | 3 - 4 | 301 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
spermatogenesis
cell differentiation
oogenesis
DNA binding
DNA-binding transcription factor activity
protein heterodimerization activity
NF-kappaB binding
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
spermatogenesis
cell differentiation
oogenesis
DNA binding
DNA-binding transcription factor activity
protein heterodimerization activity
NF-kappaB binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369091 | AK029943.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369091 | NM_001100449.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369091 | AK029943.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369091 | NM_001100449.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958866 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832955 | AX350998.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832955 | X14759.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832955 | X59251.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855363 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807865 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5924 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944610 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855819 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632268 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426794 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871004 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523810 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670985 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478117 | AK017706.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407019 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963995 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893653 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349953 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677672 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26492 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910259 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731103 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33931 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155664 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951924 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433707 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606297 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300014 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3792 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284506 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612487 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419728 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219249 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143233 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78001 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227848 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841941 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914162 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330626 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298583 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544094 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296401 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333992 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57660 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308994 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101849 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35133 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19099 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240041 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630685 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70778 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54372 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161324 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65996 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119162 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162554 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164407 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118931 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96658 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369091 | AK029943.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369091 | NM_001100449.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369091 | AK029943.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369091 | NM_001100449.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08920082 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08920057 | - | - |
Whole Mouse Genome G4122A | A_51_P248293 | NM_001100449.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P248293 | AK012135.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P369091 | AK029943.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P369091 | NM_001100449.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P369091 | AK029943.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P369091 | NM_001100449.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08920082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514255 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13929153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14094831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13924032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13800551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14038149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14104500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13799695 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13790571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13842799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514246 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13643216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14119019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13982459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13842800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514248 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14042304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13978286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31514269 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10454080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454081 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454084 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454085 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454090 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454079 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454092 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454088 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454083 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454078 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454089 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454082 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454086 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454087 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10454091 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435303_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435302_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431193_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10454077 | - | - |
Mouse Genome 430 2.0 Array | 1435303_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435302_at | - | - |
Mouse Genome 430 2.0 Array | 1435303_at | - | - |
Mouse Genome 430 2.0 Array | 1435302_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431193_at | - | - |
Mouse Genome 430 2.0 Array | 1431193_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72504_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72504_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800000908.mm.1 | TC1800000124.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800000908.mm.1 | NM_001100449.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800000908.mm.1 | NM_001100449.1 | Refseq |
Mouse Expression Set 430 | 1431193_at | - | - |
Mouse Expression Set 430 | 1431193_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435303_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435302_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431193_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435302_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431193_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435303_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 713229 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2658714 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2470074 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2658714 | - | - |
