Assay Details
Gene Symbol: Klf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033563.2 | NP_291041.2 | 1 - 2 | 490 | 74 | |
GenBank mRNA | AB221646.1 | - | 1 - 2 | 102 | 74 | |
AF327568.1 | - | 1 - 2 | 381 | 74 | ||
AF338369.1 | - | 1 - 2 | 525 | 74 | ||
AK149362.1 | - | 2 - 3 | 281 | 74 | ||
BC004700.1 | - | 2 - 3 | 255 | 3499191 | 74 | |
BC019604.1 | - | 1 - 2 | 490 | 3601534 | 74 | |
BC040794.1 | - | 2 - 3 | 255 | 3499191 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
