Assay Details
Gene Symbol: Slc11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013612.2 | NP_038640.2 | 1 - 2 | 120 | 97 | |
GenBank mRNA | AK090303.1 | - | 1 - 2 | 121 | 97 | |
AK144237.1 | - | 1 - 2 | 103 | 97 | ||
AK171393.1 | - | 1 - 2 | 101 | 97 | ||
BB873183.1 | - | 1 - 2 | 121 | 97 | ||
L13732.1 | - | 1 - 2 | 98 | 97 | ||
X75355.1 | - | 1 - 2 | 79 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK171393.1, L13732.1, NM_013612.2, X75355.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
