Assay Details
Gene Symbol: Arntl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289679.1 | NP_001276608.1 | 4 - 5 | 384 | 114 | |
NM_001289680.1 | NP_001276609.1 | 4 - 5 | 384 | 114 | ||
NM_001289681.1 | NP_001276610.1 | 4 - 5 | 384 | 114 | ||
NM_172309.2 | NP_758513.1 | 4 - 5 | 384 | 114 | ||
GenBank mRNA | AY005163.1 | - | 4 - 5 | 384 | 114 | |
AY014836.1 | - | 4 - 5 | 384 | 114 | ||
BC108965.2 | - | 4 - 5 | 384 | 40047159 | 114 | |
BC108966.1 | - | 4 - 5 | 384 | 114 | ||
DQ322241.1 | - | 4 - 5 | 384 | 114 | ||
DQ355143.1 | - | 4 - 5 | 384 | 114 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001289681.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
