Assay Details
Gene Symbol: Uba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016682.2 | NP_057891.1 | 4 - 5 | 382 | 115 | |
GenBank mRNA | AK146925.1 | - | 4 - 5 | 428 | 115 | |
AK151765.1 | - | 4 - 5 | 387 | 115 | ||
AK152415.1 | - | 4 - 5 | 406 | 115 | ||
AK152977.1 | - | 4 - 5 | 387 | 115 | ||
AK163451.1 | - | 4 - 5 | 378 | 115 | ||
AK164826.1 | - | 4 - 5 | 398 | 115 | ||
AK165321.1 | - | 4 - 5 | 404 | 115 | ||
AK166133.1 | - | 4 - 5 | 414 | 115 | ||
AK169168.1 | - | 4 - 5 | 378 | 115 | ||
BC054768.1 | - | 4 - 5 | 408 | 6415071 | 115 | |
U35833.1 | - | 4 - 5 | 383 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
