Assay Details
Gene Symbol: Raph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045513.3 | NP_001038978.3 | 11 - 12 | 1718 | 86 | |
GenBank mRNA | DV652147.1 | - | 3 - 4 | 259 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
