Assay Details
Gene Symbol: Nbeal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173444.2 | NP_775620.2 | 36 - 37 | 6090 | 151 | |
GenBank mRNA | AK143706.1 | - | 24 - 25 | 4000 | 151 | |
BC060141.1 | - | 10 - 11 | 1955 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
