Assay Details
Gene Symbol: Caprin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301351.1 | NP_001288280.1 | 12 - 13 | 1477 | 81 | |
NM_181541.4 | NP_853519.2 | 13 - 14 | 2134 | 81 | ||
GenBank mRNA | AK129460.1 | - | 12 - 13 | 1390 | 81 | |
AK140744.1 | - | 13 - 14 | 2137 | 81 | ||
BC025636.1 | - | 13 - 14 | 2090 | 81 | ||
BC125373.1 | - | 13 - 14 | 2023 | 40129888 | 81 | |
BC125375.1 | - | 13 - 14 | 2023 | 40129890 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
