Assay Details
Gene Symbol: Farsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102758.1 | 1 - 2 | 112 | 73 | ||
GenBank mRNA | AK134799.1 | - | 1 - 2 | 74 | 73 | |
AK169621.1 | - | 1 - 2 | 75 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK134799.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
