Assay Details
Gene Symbol: Farsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278075.1 | NP_001265004.1 | 10 - 11 | 956 | 87 | |
NM_011811.4 | NP_035941.2 | 10 - 11 | 956 | 87 | ||
NR_102757.1 | 10 - 11 | 858 | 87 | |||
NR_102758.1 | 9 - 10 | 900 | 87 | |||
GenBank mRNA | AF123263.1 | - | 10 - 11 | 956 | 87 | |
AK010271.1 | - | 10 - 11 | 919 | 87 | ||
AK045432.1 | - | 10 - 11 | 908 | 87 | ||
AK076607.1 | - | 10 - 11 | 918 | 87 | ||
AK134799.1 | - | 9 - 10 | 862 | 87 | ||
AK165645.1 | - | 10 - 11 | 823 | 87 | ||
AK167945.1 | - | 10 - 11 | 919 | 87 | ||
AK168923.1 | - | 10 - 11 | 900 | 87 | ||
AK169621.1 | - | 9 - 10 | 863 | 87 | ||
BC002313.1 | - | 10 - 11 | 815 | 3592805 | 87 | |
BC016428.1 | - | 10 - 11 | 923 | 4239906 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
