Assay Details
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290568.1 | NP_001277497.1 | 10 - 11 | 972 | 132 | |
NM_026319.3 | NP_080595.2 | 10 - 11 | 972 | 132 | ||
GenBank mRNA | AK011690.1 | - | 10 - 11 | 943 | 132 | |
AK014770.1 | - | 10 - 11 | 940 | 132 | ||
AK053369.1 | - | 10 - 11 | 920 | 132 | ||
BC005798.1 | - | 10 - 11 | 942 | 132 | ||
BC023760.1 | - | 10 - 11 | 904 | 132 | ||
BC049156.1 | - | 10 - 11 | 892 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
