Assay Details
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026319.3 | NP_080595.2 | 13 - 14 | 1226 | 146 | |
GenBank mRNA | AK006492.1 | - | 4 - 5 | 333 | 146 | |
BY706530.1 | - | 4 - 5 | 331 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
