Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168356.1 | NP_001161828.1 | 6 - 7 | 895 | 69 | |
NM_134253.2 | NP_599014.2 | 8 - 9 | 1123 | 69 | ||
GenBank mRNA | AK142472.1 | - | 6 - 7 | 752 | 69 | |
BC005659.1 | - | 8 - 9 | 1032 | 3588701 | 69 | |
BC006585.1 | - | 4 - 5 | 379 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
