Assay Details
Gene Symbol: Ceacam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175315.1 | NP_780524.1 | 1 - 2 | 154 | 78 | |
GenBank mRNA | AK049385.1 | - | 1 - 2 | 69 | 78 | |
BC099491.1 | - | 1 - 2 | 60 | 78 | ||
BC125404.1 | - | 1 - 2 | 154 | 40129919 | 78 | |
BC125406.1 | - | 1 - 2 | 154 | 40129921 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
