Assay Details
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205313.1 | NP_001192242.1 | 14 - 15 | 1519 | 71 | |
NM_001205314.1 | NP_001192243.1 | 14 - 15 | 1555 | 71 | ||
NM_009283.4 | NP_033309.3 | 14 - 15 | 1570 | 71 | ||
GenBank mRNA | AK018544.1 | - | 14 - 15 | 1521 | 71 | |
AK039458.1 | - | 14 - 15 | 1571 | 71 | ||
AK046517.1 | - | 14 - 15 | 1519 | 71 | ||
AK082706.1 | - | 14 - 15 | 1552 | 71 | ||
AK084855.1 | - | 14 - 15 | 1504 | 71 | ||
AK157263.1 | - | 14 - 15 | 1419 | 71 | ||
AK159887.1 | - | 14 - 15 | 1554 | 71 | ||
BC004808.1 | - | 14 - 15 | 1506 | 3587831 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
