Assay Details
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081081.2 | NP_001074550.1 | 5 - 6 | 1069 | 115 | |
NM_001113383.1 | NP_001106854.1 | 5 - 6 | 1069 | 115 | ||
GenBank mRNA | AK173039.1 | - | 5 - 6 | 1069 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
