Assay Details
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029837.2 | NP_001025008.2 | 6 - 7 | 1059 | 75 | |
NM_001164049.1 | NP_001157521.1 | 6 - 7 | 1086 | 75 | ||
NM_001164050.1 | NP_001157522.1 | 6 - 7 | 1086 | 75 | ||
NM_001164051.1 | NP_001157523.1 | 6 - 7 | 1110 | 75 | ||
NM_001164052.1 | NP_001157524.1 | 4 - 5 | 813 | 75 | ||
NM_008840.3 | NP_032866.2 | 7 - 8 | 1261 | 75 | ||
GenBank mRNA | AK040867.1 | - | 6 - 7 | 1021 | 75 | |
AK149935.1 | - | 7 - 8 | 1160 | 75 | ||
AK150649.1 | - | 6 - 7 | 987 | 75 | ||
AK151482.1 | - | 6 - 7 | 987 | 75 | ||
AK152661.1 | - | 6 - 7 | 987 | 75 | ||
AK153304.1 | - | 6 - 7 | 1021 | 75 | ||
AK155835.1 | - | 6 - 7 | 972 | 75 | ||
AK172222.1 | - | 6 - 7 | 963 | 75 | ||
BC035203.1 | - | 6 - 7 | 1076 | 4192906 | 75 | |
CT010349.1 | - | 4 - 5 | 781 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
