Assay Details
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133435.1 | NP_597679.1 | 4 - 5 | 530 | 55 | |
GenBank mRNA | AF260925.1 | - | 4 - 5 | 683 | 55 | |
AK011137.1 | - | 4 - 5 | 552 | 55 | ||
AK012194.1 | - | 4 - 5 | 764 | 55 | ||
AY679721.1 | - | 4 - 5 | 530 | 55 | ||
BC138500.1 | - | 4 - 5 | 598 | 8861520 | 55 | |
BC138502.1 | - | 4 - 5 | 598 | 8861522 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400756 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400757 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400755 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146374 | BC138500.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146374 | BC138502.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146374 | AK012194.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146374 | AK152407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146374 | AY679721.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479253 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163033 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906946 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548815 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885882 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126170 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654898 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154364 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128307 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10229 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100478 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173792 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146374 | BC138500.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146374 | BC138502.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146374 | AK012194.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146374 | AK152407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146374 | AY679721.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P91293 | AK012194.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P91293 | AK152407.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P91293 | AF260925.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393055 | AK012194.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393055 | AK152407.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393055 | AF260925.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393055 | AK011137.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400735 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05447329 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100478 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05447319 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05447311 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173792 | - | - |
Whole Mouse Genome G4122A | A_52_P91293 | AK012194.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P91293 | AK152407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P91293 | AF260925.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393055 | AK012194.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393055 | AK152407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393055 | AF260925.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P393055 | AK011137.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100478 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173792 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03599379 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03599364 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400755 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400756 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400739 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03935868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400719 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400727 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03674568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03222878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03674566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03959158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03213441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03374690 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03599364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03935866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400729 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23400733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03213438 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10518681 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518685 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518683 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518680 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518684 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518682 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429819_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425773_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10518679 | - | - |
Mouse Genome 430 2.0 Array | 1429819_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1425773_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1429819_at | - | - |
Mouse Genome 430 2.0 Array | 1425773_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66454_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66454_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400033585.mm.1 | TC0400004042.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400033585.mm.1 | NM_133435.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400033585.mm.1 | NM_133435.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400033584.mm.1 | TC0400004042.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425773_s_at | - | - |
Mouse Expression Set 430 | 1425773_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1425773_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1425773_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1429819_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425773_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1429819_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425773_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2502596 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2502596 | - | - |
