Assay Details
Gene Symbol: Scp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011327.4 | NP_035457.1 | 9 - 10 | 924 | 94 | |
GenBank mRNA | AK004860.2 | - | 9 - 10 | 854 | 94 | |
BC018384.1 | - | 9 - 10 | 879 | 3710340 | 94 | |
BC034613.1 | - | 9 - 10 | 845 | 5123611 | 94 | |
M91458.1 | - | 9 - 10 | 839 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
