Assay Details
Gene Symbol: Rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153793.2 | NP_722488.2 | 4 - 5 | 724 | 122 | |
GenBank mRNA | AK044097.1 | - | 4 - 5 | 687 | 122 | |
BC038500.1 | - | 4 - 5 | 667 | 4481989 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31631095 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31631093 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31631094 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093168 | BC057367.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093168 | AK154732.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093168 | BC006858.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093168 | NM_175684.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093168 | BC038500.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047106 | BC057367.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047106 | AK154732.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047106 | BC006858.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047106 | NM_175684.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047106 | AK044097.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369869 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859873 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133440 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047106 | BC057367.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047106 | AK154732.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047106 | BC006858.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047106 | NM_175684.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047106 | AK044097.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31631093 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133440 | - | - |
Whole Mouse Genome G4122A | A_52_P268880 | BC057367.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P268880 | AK154732.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P268880 | BC006858.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P268880 | NM_175684.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P268880 | AK044097.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133440 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31631095 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31631094 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31631093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31631177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631097 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31631109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13689632 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10455203 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455201 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455202 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455205 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455208 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455207 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455209 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455200 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455206 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455204 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434147_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10455199 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10458478 | - | - |
Mouse Genome 430 2.0 Array | 1427155_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1434147_at | - | - |
Mouse Genome 430 2.0 Array | 1434147_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427155_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225392_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225392_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800002922.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002922.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002922.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002926.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002926.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002926.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002929.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002929.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002929.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002918.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002918.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002918.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002927.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002917.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002917.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002917.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002921.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002921.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002921.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002923.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002923.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002923.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002924.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002920.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002920.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002920.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002925.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002925.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002925.mm.1 | NM_153793.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002928.mm.1 | TC1800000385.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800002928.mm.1 | NM_153793.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002928.mm.1 | NM_153793.2 | Refseq |
Mouse Expression Set 430 | 1427155_at | - | - |
Mouse Expression Set 430 | 1434147_at | - | - |
Mouse Expression Set 430 | 1434147_PM_at | - | - |
Mouse Expression Set 430 | 1427155_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427155_at | - | - |
Mouse Genome 430A 2.0 Array | 1427155_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1434147_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1434147_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434147_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434147_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5860746 | - | - |
MouseWG-6 v2 Expression BeadChip | 770487 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 770487 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5860746 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000770487 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005860746 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671794 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2671794 | - | - |
mouse-6 v1.1 (Illumina) | scl52045.2_4-S | - | - |
mouse-6 v1.1 (Illumina) | scl00225392.1_86-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5860746 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 770487 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00225392.1_86-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52045.2_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860746 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770487 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2671794 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2916437 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2916437 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671794 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227065 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl52045.2_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00225392.1_86-S | - | - |
