Assay Details
Gene Symbol: Tars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163618.1 | NP_001157090.1 | 10 - 11 | 1447 | 61 | |
NM_001163619.1 | NP_001157091.1 | 10 - 11 | 1309 | 61 | ||
NM_027931.3 | NP_082207.2 | 12 - 13 | 1690 | 61 | ||
GenBank mRNA | AK011540.1 | - | 12 - 13 | 1576 | 61 | |
AK142685.1 | - | 12 - 13 | 1585 | 61 | ||
AK164589.1 | - | 10 - 11 | 1216 | 61 | ||
AK164825.1 | - | 10 - 11 | 1088 | 61 | ||
AK168075.1 | - | 10 - 11 | 1341 | 61 | ||
AK204901.1 | - | 1 - 2 | 48 | 61 | ||
BC067392.1 | - | 6 - 7 | 2798 | 61 | ||
BC147501.1 | - | 12 - 13 | 1574 | 9056383 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
