Assay Details
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134791.2 | NP_001128263.1 | 9 - 10 | 832 | 70 | |
NM_133885.4 | NP_598646.4 | 12 - 13 | 1065 | 70 | ||
NM_173350.3 | NP_775485.2 | 10 - 11 | 871 | 70 | ||
GenBank mRNA | AK035658.1 | - | 10 - 11 | 820 | 70 | |
AK131984.1 | - | 12 - 13 | 1036 | 70 | ||
AK172010.1 | - | 10 - 11 | 858 | 70 | ||
AK184542.1 | - | 1 - 2 | 70 | 70 | ||
AK199715.1 | - | 1 - 2 | 70 | 70 | ||
BC021507.1 | - | 8 - 9 | 693 | 5347121 | 70 | |
BC023759.1 | - | 12 - 13 | 1020 | 70 | ||
BC026927.1 | - | 10 - 11 | 754 | 5134780 | 70 | |
BC089163.1 | - | 9 - 10 | 773 | 5367603 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
