Assay Details
Gene Symbol: Ablim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164491.1 | NP_001157963.1 | 10 - 11 | 1146 | 75 | |
NM_198649.3 | NP_941051.2 | 9 - 10 | 1465 | 75 | ||
GenBank mRNA | AK086538.1 | - | 9 - 10 | 1245 | 75 | |
AK173040.1 | - | 9 - 10 | 1023 | 75 | ||
BC060275.1 | - | 10 - 11 | 1117 | 6822626 | 75 | |
BC094229.1 | - | 9 - 10 | 1439 | 30534236 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
